Anti-SET7/9 Rabbit Polyclonal Antibody

CAYM13780-500EA 532 EUR
CAYM13780-500
Anti-SET7/9 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SET7/9
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR