Anti-CASP14 Mouse Monoclonal Antibody [clone: 70A1426]

Toimittaja: Cayman Chemical
CAYM13916-1EA 580 EUR
CAYM13916-1
Anti-CASP14 Mouse Monoclonal Antibody [clone: 70A1426]
Vasta-aineet
Type: Primary
Antigen: CASP14
Clonality: Monoclonal
Clone: 70A1426
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR