Anti-COX-2 Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM160106-1EA 442 EUR
CAYM160106-1
Anti-COX-2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: COX-2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Guinea Pig, Monkey, Mouse, Sheep, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR