Anti-COX-1 Rabbit Polyclonal Antibody

CAYM160109-1EA 532 EUR
CAYM160109-1
Anti-COX-1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: COX-1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR