Anti-PGIS Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM160640-1EA 442 EUR
CAYM160640-1
Anti-PGIS Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: PGIS
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR