Anti-ALDH1 Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM160720-1EA 442 EUR
CAYM160720-1
Anti-ALDH1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ALDH1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Sheep, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR