Anti-iNOS Rabbit Polyclonal Antibody

Toimittaja: Cayman Chemical
CAYM160862-1EA 442 EUR
CAYM160862-1
Anti-iNOS Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: iNOS
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: most Mammal species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR