Anti-HA Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))

Toimittaja: Cayman Chemical
CAYM162200-1EA 442 EUR
CAYM162200-1
Anti-HA Rabbit Polyclonal Antibody (KLH (Keyhole Limpet Hemocyanin))
Vasta-aineet
Type: Tag
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: KLH (Keyhole Limpet Hemocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR