Anti-Vtg Mouse Monoclonal Antibody [clone: ND-5F8]

Toimittaja: Cayman Chemical
CAYM170195-100EA 402 EUR
CAYM170195-100 CAYM170195-500
Anti-Vtg Mouse Monoclonal Antibody [clone: ND-5F8]
Vasta-aineet
Type: Primary
Antigen: Vtg
Clonality: Monoclonal
Clone: ND-5F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: common Carp, Halibut, killifish, Sheepshead Minnow, Tilapia, White Perch, Wrasse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR