Anti-LYN Mouse Monoclonal Antibody [clone: 1174CT22.3.1.1]

Toimittaja: Abgent
ABGEAM2258AEA 360 EUR
ABGEAM2258A
Anti-LYN Mouse Monoclonal Antibody [clone: 1174CT22.3.1.1]
Vasta-aineet
Type: Primary
Antigen: LYN
Clonality: Monoclonal
Clone: 1174CT22.3.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR