Anti-MYH7 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP17633AEA 350 EUR
ABGEAP17633A
Anti-MYH7 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MYH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR