Anti-JAK1 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP20699AEA 348 EUR
ABGEAP20699A
Anti-JAK1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: JAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR