Anti-SP2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP13121BEA 348 EUR
ABGEAP13121B
Anti-SP2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR