Anti-ECSCR Rabbit Polyclonal Antibody

Toimittaja: Abgent
ABGEAP18022BEA 348 EUR
ABGEAP18022B
Anti-ECSCR Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ECSCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR