Anti-IgG Rabbit Monoclonal Antibody (HRP (Horseradish Peroxidase))

Toimittaja: Abnova
ABNOPAB10668EA 356 EUR
ABNOPAB10668
Anti-IgG Rabbit Monoclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Rabbit anti-horse IgG recognizes horse IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Horse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR