Anti-ZFYVE20 Goat Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB6458EA 440 EUR
ABNOPAB6458
Anti-ZFYVE20 Goat Polyclonal Antibody
Vasta-aineet
Goat polyclonal antibody raised against synthetic peptide of ZFYVE20.

Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1.5 ?g/ml

Type: Primary
Antigen: ZFYVE20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR