Anti-Bordetella pertussis Mouse Monoclonal Antibody [clone: BDI500]

Toimittaja: Abnova
ABNOMAB3897EA 1210 EUR
ABNOMAB3897
Anti-Bordetella pertussis Mouse Monoclonal Antibody [clone: BDI500]
Mouse monoclonal antibody raised against Bordetella pertussis.

Type: Primary
Antigen: Bordetella pertussis
Clonality: Monoclonal
Clone: BDI500
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR