Anti-RSPO3 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB22374EA 545 EUR
ABNOPAB22374
Anti-RSPO3 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant RSPO3.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: RSPO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR