Anti-TRPV1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB0690EA 338 EUR
ABNOPAB0690
Anti-TRPV1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic peptide of TRPV1.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:2000

Type: Primary
Antigen: TRPV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR