Anti-STC1 Mouse Monoclonal Antibody [clone: 4C11]

Toimittaja: Abnova
ABNOMAB0034-M01EA 344 EUR
ABNOMAB0034-M01
Anti-STC1 Mouse Monoclonal Antibody [clone: 4C11]
Vasta-aineet
Mouse monoclonal antibody raised against stanniocalcin-1.

Type: Primary
Antigen: STC1
Clonality: Monoclonal
Clone: 4C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR