Anti-GATA4 Mouse Monoclonal Antibody [clone: 6H10]

Toimittaja: Abnova
ABNOMAB10664EA 440 EUR
ABNOMAB10664
Anti-GATA4 Mouse Monoclonal Antibody [clone: 6H10]
Vasta-aineet
Mouse monoclonal antibody raised against partial recombinant GATA4.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: GATA4
Clonality: Monoclonal
Clone: 6H10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR