Anti-NDP Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28650EA 518 EUR
ABNOPAB28650
Anti-NDP Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant NDP.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: NDP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR