Anti-MDP1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28644EA 518 EUR
ABNOPAB28644
Anti-MDP1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant MDP-1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:100-1:250

Type: Primary
Antigen: MDP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR