Anti-SAGE1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28634EA 518 EUR
ABNOPAB28634
Anti-SAGE1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant SAGE1.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: SAGE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR