Anti-LAX1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28667EA 518 EUR
ABNOPAB28667
Anti-LAX1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant LAX1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:100-1:250

Type: Primary
Antigen: LAX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR