Anti-RAB28 Mouse Polyclonal Antibody
Toimittaja: Abnova
ABNOH00009364-B03PEA 394 EUR
ABNOH00009364-B03P
Anti-RAB28 Mouse Polyclonal Antibody
Vasta-aineet
Mouse polyclonal antibody raised against a full-length human RAB28 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: RAB28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now