Anti-CD302 Mouse Monoclonal Antibody [clone: 8G7]
Toimittaja: Abnova
ABNOH00009936-M01EA 394 EUR
ABNOH00009936-M01
Anti-CD302 Mouse Monoclonal Antibody [clone: 8G7]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant CD302.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CD302
Clonality: Monoclonal
Clone: 8G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now