Anti-FRAT1 Mouse Monoclonal Antibody [clone: 7C9]
Toimittaja: Abnova
ABNOH00010023-M01EA 394 EUR
ABNOH00010023-M01
Anti-FRAT1 Mouse Monoclonal Antibody [clone: 7C9]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant FRAT1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: FRAT1
Clonality: Monoclonal
Clone: 7C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now