Anti-ITPR1 Mouse Monoclonal Antibody (ATTO 594) [clone: S24-18]

Toimittaja: Abnova
ABNOMAB11678EA 545 EUR
ABNOMAB11678
Anti-ITPR1 Mouse Monoclonal Antibody (ATTO 594) [clone: S24-18]
Vasta-aineet
Mouse monoclonal antibody raised against partial synthetic protein of Itpr1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: ITPR1
Clonality: Monoclonal
Clone: S24-18
Conjugation: ATTO 594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR