Anti-SCNN1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Toimittaja: Abnova
ABNOPAB28779EA 545 EUR
ABNOPAB28779
Anti-SCNN1A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant Rat Scnn1a.
Recommended Dilutions: Immunofluorescence: 1:400; Immunohistochemistry Western Blot: 1:1000
Type:
Antigen: SCNN1A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now