Anti-SCNN1G Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28802EA 440 EUR
ABNOPAB28802
Anti-SCNN1G Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant Rat Scnn1g.

Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SCNN1G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR