Anti-AQP1 Rabbit Polyclonal Antibody (ATTO 488)

Toimittaja: Abnova
ABNOPAB28840EA 772 EUR
ABNOPAB28840
Anti-AQP1 Rabbit Polyclonal Antibody (ATTO 488)
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant Rat Aqp1.

Recommended Dilutions: Immunofluorescence: 1:200-400; Western Blot: 1:1000-4000

Type:
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR