Anti-AQP1 Rabbit Polyclonal Antibody (Biotin)
Toimittaja: Abnova
ABNOPAB28849EA 772 EUR
ABNOPAB28849
Anti-AQP1 Rabbit Polyclonal Antibody (Biotin)
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant Rat Aqp1.
Recommended Dilutions: Immunofluorescence: 1:200-400; Western Blot: 1:1000-4000
Type:
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now