Anti-MIF4GD Mouse Polyclonal Antibody

Toimittaja: Abnova
ABNOH00057409-B01PEA 394 EUR
ABNOH00057409-B01P
Anti-MIF4GD Mouse Polyclonal Antibody
Vasta-aineet
Mouse polyclonal antibody raised against a full-length human MIF4GD protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: MIF4GD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR