Anti-GPR135 Mouse Monoclonal Antibody [clone: 1H5]

Toimittaja: Abnova
ABNOH00064582-M01EA 394 EUR
ABNOH00064582-M01
Anti-GPR135 Mouse Monoclonal Antibody [clone: 1H5]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant GPR135.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: GPR135
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR