Anti-HSPA1A Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 2A4]
Toimittaja: Abnova
ABNOMAB11755EA 545 EUR
ABNOMAB11755
Anti-HSPA1A Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 2A4]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant HSPA1A.
Recommended Dilutions: Western Blot: 1:5000; Immunocytochemistry: 1:100; Immunofluorescence: 1:100; Immunohistochemistry: 1:100
Type: Primary
Antigen: HSPA1A
Clonality: Monoclonal
Clone: 2A4
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now