Anti-AQP3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Toimittaja: Abnova
ABNOPAB28883EA 545 EUR
ABNOPAB28883
Anti-AQP3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant Rat Aqp3.

Recommended Dilutions: Immunofluorescence: 1:200-400; Western Blot: 1:1000-4000

Type:
Antigen: AQP3
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR