Anti-SPATA19 Mouse Monoclonal Antibody [clone: 4A4]

Toimittaja: Abnova
ABNOH00219938-M01EA 394 EUR
ABNOH00219938-M01
Anti-SPATA19 Mouse Monoclonal Antibody [clone: 4A4]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant SPATA19.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SPATA19
Clonality: Monoclonal
Clone: 4A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR