Anti-NANOS1 Mouse Monoclonal Antibody [clone: 5F12]

Toimittaja: Abnova
ABNOH00340719-M01EA 394 EUR
ABNOH00340719-M01
Anti-NANOS1 Mouse Monoclonal Antibody [clone: 5F12]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant NANOS1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: NANOS1
Clonality: Monoclonal
Clone: 5F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR