Anti-TXNDC5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2E7/7]
Toimittaja: Abnova
ABNOMAB11792EA 545 EUR
ABNOMAB11792
Anti-TXNDC5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2E7/7]
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptide of TXNDC5.
Recommended Dilutions: Western Blot: 1:200
Type: Primary
Antigen: TXNDC5
Clonality: Monoclonal
Clone: 2E7/7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now