Anti-AQP4 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Toimittaja: Abnova
ABNOPAB28901EA 545 EUR
ABNOPAB28901
Anti-AQP4 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant Rat Aqp4.

Recommended Dilutions: Immunofluorescence: 1:400; Western Blot: 1:1000-4000

Type:
Antigen: AQP4
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR