Anti-APP Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28910EA 643 EUR
ABNOPAB28910
Anti-APP Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody against amyloid fibrils and fibrillar oligomers.

  • Recommended Dilutions: Dot Blot: 1:1000

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR