Anti-APP Rabbit Polyclonal Antibody (ATTO 488)
Toimittaja: Abnova
ABNOPAB28912EA 545 EUR
ABNOPAB28912
Anti-APP Rabbit Polyclonal Antibody (ATTO 488)
Vasta-aineet
Rabbit polyclonal antibody against amyloid fibrils and fibrillar oligomers.
Recommended Dilutions: Dot Blot: 1:1000
Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now