Anti-APP Rabbit Polyclonal Antibody (ATTO 594)

Toimittaja: Abnova
ABNOPAB28914EA 545 EUR
ABNOPAB28914
Anti-APP Rabbit Polyclonal Antibody (ATTO 594)
Vasta-aineet
Rabbit polyclonal antibody against amyloid fibrils and fibrillar oligomers.

Recommended Dilutions: Dot Blot: 1:1000

Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: ATTO 594
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR