Anti-KCNB2 Mouse Monoclonal Antibody (ATTO 594) [clone: S37-89]

Toimittaja: Abnova
ABNOMAB11840EA 545 EUR
ABNOMAB11840
Anti-KCNB2 Mouse Monoclonal Antibody (ATTO 594) [clone: S37-89]
Vasta-aineet
Mouse monoclonal antibody raised against partial recombinant Kcnb2.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: KCNB2
Clonality: Monoclonal
Clone: S37-89
Conjugation: ATTO 594
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR