Anti-KCNAB1 Mouse Monoclonal Antibody (ATTO 488) [clone: S47-42]
Toimittaja: Abnova
ABNOMAB11856EA 545 EUR
ABNOMAB11856
Anti-KCNAB1 Mouse Monoclonal Antibody (ATTO 488) [clone: S47-42]
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptide of Kcnab1.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: KCNAB1
Clonality: Monoclonal
Clone: S47-42
Conjugation: ATTO 488
Epitope: N-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now