Anti-APP Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Toimittaja: Abnova
ABNOPAB28944EA 545 EUR
ABNOPAB28944
Anti-APP Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Vasta-aineet
Rabbit polyclonal antibody against amyloid oligomers.

Recommended Dilutions: Immunohistochemistry: 1:1000-10000; Immunoprecipitation: 1:1000; Western Blot

Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR