Anti-KCNAB1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S47-42]

Toimittaja: Abnova
ABNOMAB11866EA 545 EUR
ABNOMAB11866
Anti-KCNAB1 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S47-42]
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptide of Kcnab1.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: KCNAB1
Clonality: Monoclonal
Clone: S47-42
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: N-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR