Anti-SH3GL1 Mouse Monoclonal Antibody (ATTO 488) [clone: S51-1]

Toimittaja: Abnova
ABNOMAB11874EA 545 EUR
ABNOMAB11874
Anti-SH3GL1 Mouse Monoclonal Antibody (ATTO 488) [clone: S51-1]
Vasta-aineet
Mouse monoclonal antibody raised aginst synthetic peptide of Sh3gl1.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml

Type: Primary
Antigen: SH3GL1
Clonality: Monoclonal
Clone: S51-1
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR