Anti-SH3GL1 Mouse Monoclonal Antibody (Biotin) [clone: S51-1]
Toimittaja: Abnova
ABNOMAB11883EA 545 EUR
ABNOMAB11883
Anti-SH3GL1 Mouse Monoclonal Antibody (Biotin) [clone: S51-1]
Vasta-aineet
Mouse monoclonal antibody raised aginst synthetic peptide of Sh3gl1.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Immunohistochemistry: 0.1-1 ?g/ml
Type: Primary
Antigen: SH3GL1
Clonality: Monoclonal
Clone: S51-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now